Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011082.3 | NP_035212.2 | 8 - 9 | 2211 | 60 | |
GenBank mRNA | AK128944.1 | - | 8 - 9 | 2211 | 60 | |
AK142656.1 | - | 8 - 9 | 2213 | 60 | ||
BC013556.1 | - | 8 - 9 | 2144 | 3601095 | 60 | |
U06431.1 | - | 8 - 9 | 2115 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |