Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011126.3 | NP_035256.2 | 6 - 7 | 792 | 67 | |
GenBank mRNA | BC054375.1 | - | 6 - 7 | 773 | 6314015 | 67 |
U69172.1 | - | 6 - 7 | 788 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |