Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtfp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026443.4 | NP_080719.2 | 2 - 3 | 346 | 63 | |
GenBank mRNA | AK006148.1 | - | 2 - 3 | 267 | 63 | |
AK010971.1 | - | 2 - 3 | 303 | 63 | ||
AK012058.1 | - | 2 - 3 | 313 | 63 | ||
AK078381.1 | - | 2 - 3 | 347 | 63 | ||
AK167990.1 | - | 2 - 3 | 213 | 63 | ||
BC019205.1 | - | 2 - 3 | 356 | 63 | ||
BC058737.1 | - | 2 - 3 | 296 | 5400435 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |