Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027000.4 | NP_081276.2 | 3 - 4 | 333 | 108 | |
GenBank mRNA | AF325354.1 | - | 3 - 4 | 317 | 108 | |
AF348208.1 | - | 3 - 4 | 340 | 108 | ||
AK011580.1 | - | 3 - 4 | 344 | 108 | ||
AK141852.1 | - | 3 - 4 | 343 | 108 | ||
AK163372.1 | - | 2 - 3 | 140 | 108 | ||
AK164330.1 | - | 3 - 4 | 334 | 108 | ||
AK166711.1 | - | 3 - 4 | 339 | 108 | ||
BC005514.1 | - | 3 - 4 | 314 | 108 | ||
BC025431.1 | - | 3 - 4 | 338 | 5007174 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |