Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027211.2 | NP_081487.1 | 766 | 69 | ||
GenBank mRNA | AJ306451.1 | - | 766 | 69 | ||
AK007692.1 | - | 1 - 2 | 147 | 69 | ||
BC013521.1 | - | 736 | 4236501 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |