Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029420.4 | NP_083696.2 | 4 - 5 | 880 | 60 | |
NR_033446.1 | 3 - 4 | 828 | 60 | |||
GenBank mRNA | AK010748.1 | - | 4 - 5 | 880 | 60 | |
AK014753.1 | - | 3 - 4 | 1002 | 60 | ||
AK049139.1 | - | 4 - 5 | 881 | 60 | ||
AK053901.1 | - | 3 - 4 | 769 | 60 | ||
BC055909.1 | - | 4 - 5 | 831 | 60 | ||
BC145468.1 | - | 3 - 4 | 692 | 60 | ||
BC145688.1 | - | 4 - 5 | 745 | 40130991 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |