Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hook2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167991.1 | NP_001161463.1 | 16 - 17 | 1712 | 76 | |
NM_133255.2 | NP_573518.2 | 16 - 17 | 1712 | 76 | ||
GenBank mRNA | AK042048.1 | - | 16 - 17 | 1700 | 76 | |
AK136069.1 | - | 17 - 18 | 1634 | 76 | ||
AK145768.1 | - | 16 - 17 | 1680 | 76 | ||
AK166954.1 | - | 16 - 17 | 1692 | 76 | ||
AK169073.1 | - | 16 - 17 | 1680 | 76 | ||
BC002226.1 | - | 8 - 9 | 921 | 76 | ||
BC019486.1 | - | 16 - 17 | 1688 | 4211990 | 76 | |
BC026609.1 | - | 8 - 9 | 872 | 76 | ||
BC029967.1 | - | 16 - 17 | 1653 | 76 | ||
BC055881.1 | - | 16 - 17 | 1658 | 3590336 | 76 | |
BC080744.1 | - | 16 - 17 | 1702 | 30471369 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |