Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133701.2 | NP_598462.1 | 18 - 19 | 2574 | 92 | |
GenBank mRNA | AK004462.1 | - | 3 - 4 | 274 | 92 | |
AK081998.1 | - | 18 - 19 | 2509 | 92 | ||
AK145461.1 | - | 18 - 19 | 2577 | 92 | ||
AK166890.1 | - | 18 - 19 | 2526 | 92 | ||
BC005801.1 | - | 8 - 9 | 1090 | 92 | ||
BC014869.1 | - | 18 - 19 | 2501 | 3602158 | 92 | |
BC023691.1 | - | 18 - 19 | 2503 | 5344332 | 92 | |
CJ228927.1 | - | 2 - 3 | 185 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |