Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134102.4 | NP_598863.3 | 1 - 2 | 344 | 65 | |
GenBank mRNA | AF063498.1 | - | 1 - 2 | 345 | 65 | |
BC003470.1 | - | 1 - 2 | 87 | 2649848 | 65 | |
BC030670.1 | - | 1 - 2 | 343 | 1247206 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |