Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253843.1 | NP_001240772.1 | 14 - 15 | 1591 | 62 | |
NM_001253844.1 | NP_001240773.1 | 12 - 13 | 1184 | 62 | ||
NM_134129.4 | NP_598890.1 | 14 - 15 | 1534 | 62 | ||
GenBank mRNA | AB222602.1 | - | 14 - 15 | 1376 | 62 | |
AF251503.1 | - | 14 - 15 | 1499 | 62 | ||
AK047693.1 | - | 14 - 15 | 1409 | 62 | ||
AK076998.1 | - | 12 - 13 | 1185 | 62 | ||
AK088769.1 | - | 14 - 15 | 1519 | 62 | ||
AK164681.1 | - | 14 - 15 | 1522 | 62 | ||
BC004070.1 | - | 14 - 15 | 1473 | 3591050 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |