Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153507.2 | NP_705727.1 | 12 - 13 | 1326 | 131 | |
GenBank mRNA | AK160238.1 | - | 12 - 13 | 1316 | 131 | |
BC022128.1 | - | 12 - 13 | 1287 | 5097527 | 131 | |
BC031801.1 | - | 12 - 13 | 1326 | 3985959 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |