Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153574.2 | NP_705802.1 | 14 - 15 | 1733 | 63 | |
GenBank mRNA | AJ510136.1 | - | 14 - 15 | 1719 | 63 | |
AK036291.1 | - | 7 - 8 | 940 | 63 | ||
AK052421.1 | - | 14 - 15 | 1674 | 63 | ||
BC005554.1 | - | 14 - 15 | 1638 | 63 | ||
BC054053.1 | - | 14 - 15 | 1707 | 63 | ||
BC060978.1 | - | 14 - 15 | 1670 | 63 | ||
BC080861.1 | - | 14 - 15 | 1733 | 30357292 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |