Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flywch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153791.2 | NP_722486.2 | 3 - 4 | 951 | 69 | |
GenBank mRNA | AK034869.1 | - | 2 - 3 | 545 | 69 | |
AK052314.1 | - | 1 - 2 | 163 | 69 | ||
AK129391.1 | - | 1 - 2 | 109 | 69 | ||
AK168389.1 | - | 3 - 4 | 886 | 69 | ||
BC038031.1 | - | 2 - 3 | 1243 | 4021754 | 69 | |
CA321280.1 | - | 2 - 3 | 612 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |