Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scgb3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170727.2 | NP_733923.1 | 2 - 3 | 228 | 64 | |
GenBank mRNA | AF313457.1 | - | 2 - 3 | 213 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Inventoried Probe spans exons |