Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172049.2 | NP_742046.2 | 2 - 3 | 240 | 69 | |
GenBank mRNA | AK133493.1 | - | 2 - 3 | 240 | 69 | |
AK160817.1 | - | 2 - 3 | 232 | 69 | ||
AK166323.1 | - | 2 - 3 | 257 | 69 | ||
AK170496.1 | - | 2 - 3 | 223 | 69 | ||
BC024093.1 | - | 2 - 3 | 212 | 5116657 | 69 | |
BY013255.1 | - | 2 - 3 | 241 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |