Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P103818 |
BC054474.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P103818 |
X87257.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P103818 |
AK132354.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P103818 |
NM_007922.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1954986 |
AK165598.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
170261 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
265200 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
42036 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
552669 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
568058 |
AK044361.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
414040 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
500314 |
AK006319.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
500314 |
AK006788.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
500314 |
BC049569.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
500314 |
BC059721.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
500314 |
NM_026096.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
954512 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
492491 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
381333 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
114821 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
777481 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
269157 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
369572 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
69592 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
61352 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P155976 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P108566 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P103818 |
BC054474.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P103818 |
X87257.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P103818 |
AK132354.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P103818 |
NM_007922.4 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1954986 |
AK165598.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P1028571 |
AK082260.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290807 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290800 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290796 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290810 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290801 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290725 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290740 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290799 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290792 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290788 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290741 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290737 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290805 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290728 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290742 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290727 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290790 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290733 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290802 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290797 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290787 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290795 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290809 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290739 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290791 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290803 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290736 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290798 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32290730 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P155976 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09250462 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09250477 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09250466 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P108566 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P1028571 |
AK082260.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P103818 |
BC054474.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P103818 |
X87257.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P103818 |
AK132354.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P103818 |
NM_007922.4 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P155976 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P108566 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14847321 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14847317 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14847323 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15042246 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14847314 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14847318 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290656 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290736 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290752 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290737 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290763 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290724 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290702 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290803 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290773 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290841 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290711 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290668 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290838 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290766 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290657 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290665 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290792 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290659 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290797 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290829 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290782 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290705 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290749 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290666 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290756 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290652 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290722 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290695 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290730 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290675 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290741 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290685 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290846 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290733 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290833 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290835 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290786 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290800 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290791 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290776 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290761 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290709 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290714 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290785 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290684 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290751 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290650 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290667 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290720 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290728 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290670 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290742 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290655 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290694 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290653 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290739 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290727 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290805 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290779 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290682 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290651 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290799 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290676 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290809 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290746 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290783 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290774 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290718 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290795 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290660 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290759 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290690 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290671 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290707 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290713 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290839 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290831 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290788 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290686 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290649 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290778 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290689 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290692 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290673 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290807 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290696 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290715 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290680 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290801 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290769 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290647 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290708 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290810 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290781 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290771 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290664 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290772 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290698 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290743 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290790 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32290764 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290723 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290743 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290752 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290730 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290784 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290770 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14585857 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290720 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290782 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290737 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290790 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290778 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290736 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290809 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290733 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15050554 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15042252 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15042247 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290797 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14708352 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290805 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14847314 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290721 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290746 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290725 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290735 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290728 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14984271 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290800 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15050576 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290741 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290793 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290779 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290751 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290781 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14806117 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290771 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290810 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290768 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14864872 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290788 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290776 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15050579 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290807 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290722 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14903386 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290715 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290727 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15001131 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290787 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290799 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290792 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32290739 |
- |
- |