Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ercc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007948.2 | NP_031974.2 | 7 - 8 | 839 | 62 | |
GenBank mRNA | AK088271.1 | - | 7 - 8 | 811 | 62 | |
AK136733.1 | - | 7 - 8 | 799 | 62 | ||
BC011224.1 | - | 7 - 8 | 809 | 4039274 | 62 | |
BE949768.1 | - | 3 - 4 | 62 | |||
X07414.1 | - | 7 - 8 | 804 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |