Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007955.3 | NP_031981.3 | 25 - 26 | 4607 | 68 | |
GenBank mRNA | AF300701.1 | - | 24 - 25 | 4084 | 68 | |
AK156112.1 | - | 25 - 26 | 4610 | 68 | ||
U36488.1 | - | 21 - 22 | 3769 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Detect Genomic DNA Mus musculus Probe spans exons |