Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ext2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010163.3 | NP_034293.2 | 2 - 3 | 751 | 114 | |
GenBank mRNA | AK082572.1 | - | 2 - 3 | 733 | 114 | |
AK148090.1 | - | 2 - 3 | 740 | 114 | ||
AK148369.1 | - | 1 - 2 | 512 | 114 | ||
AK150155.1 | - | 2 - 3 | 743 | 114 | ||
AK152265.1 | - | 851 | 114 | |||
AK152840.1 | - | 853 | 114 | |||
AK164342.1 | - | 2 - 3 | 737 | 114 | ||
BC006597.1 | - | 2 - 3 | 724 | 3965079 | 114 | |
U72141.1 | - | 2 - 3 | 730 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |