Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013515.2 | NP_038543.1 | 4 - 5 | 411 | 62 | |
GenBank mRNA | AF093620.1 | - | 4 - 5 | 362 | 62 | |
AK077435.1 | - | 4 - 5 | 379 | 62 | ||
AK139242.1 | - | 4 - 5 | 379 | 62 | ||
AK143724.1 | - | 4 - 5 | 379 | 62 | ||
AK148975.1 | - | 4 - 5 | 380 | 62 | ||
AK149689.1 | - | 4 - 5 | 379 | 62 | ||
AK149821.1 | - | 4 - 5 | 379 | 62 | ||
AK149991.1 | - | 4 - 5 | 379 | 62 | ||
AK151129.1 | - | 4 - 5 | 378 | 62 | ||
BC003789.1 | - | 4 - 5 | 374 | 3593052 | 62 | |
U17297.1 | - | 4 - 5 | 411 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |