Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28882596 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P464822 |
BC089600.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P464822 |
NM_015787.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P464822 |
BC089600.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P464822 |
NM_015787.3 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
496663 |
AK035008.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
496663 |
XM_001473996.1 |
refseq_mrna_mouse_XM |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P146512 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P136403 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P464822 |
BC089600.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P464822 |
NM_015787.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P464822 |
BC089600.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P464822 |
NM_015787.3 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P464822 |
BC089600.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P464822 |
NM_015787.3 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P464822 |
BC089600.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P464822 |
NM_015787.3 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28882620 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28882622 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28882581 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28882611 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28882624 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28882596 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28882628 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28882607 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28882626 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28882630 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28882597 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28882599 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28882613 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28882603 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28882604 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28882601 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28882610 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28882618 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28882617 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28882621 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28882615 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28882577 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28882578 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P464822 |
BC089600.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P464822 |
NM_015787.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P464822 |
BC089600.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P464822 |
NM_015787.3 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P122574 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10979452 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10979441 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10979429 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10979434 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10979433 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28882596 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882538 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882615 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882521 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882542 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882552 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882575 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882525 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882548 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882617 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882565 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882642 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882558 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882645 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882529 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882519 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882568 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882577 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882528 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882546 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882613 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882636 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882596 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882533 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882608 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882611 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882639 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882634 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882517 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882567 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882601 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882646 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882610 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882637 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882544 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882555 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882557 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882626 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882572 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882560 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882556 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882574 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882582 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882523 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882607 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882651 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882531 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882618 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882635 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882641 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882621 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882599 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882649 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882622 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882576 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882604 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882630 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882561 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882578 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882537 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882583 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28882647 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28882601 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28882634 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28882596 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28882613 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28882629 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28882608 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28882635 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28882630 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28882622 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28882637 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28882599 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10999378 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28882610 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28882621 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28882607 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28882615 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28882620 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28882636 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28882611 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28882618 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28882624 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28882626 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28882604 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28882628 |
- |
- |