Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285423.1 | NP_001272352.1 | 3 - 4 | 387 | 93 | |
NM_016903.5 | NP_058599.1 | 3 - 4 | 254 | 93 | ||
GenBank mRNA | AB025408.1 | - | 3 - 4 | 229 | 93 | |
AK007527.1 | - | 3 - 4 | 242 | 93 | ||
AK010683.1 | - | 3 - 4 | 259 | 93 | ||
AK021209.1 | - | 3 - 4 | 182 | 93 | ||
AK079131.1 | - | 3 - 4 | 207 | 93 | ||
AK088115.1 | - | 3 - 4 | 302 | 93 | ||
AK163074.1 | - | 3 - 4 | 182 | 93 | ||
BC046766.1 | - | 3 - 4 | 179 | 5694094 | 93 | |
BC096644.1 | - | 3 - 4 | 194 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |