Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021292.2 | NP_067267.2 | 9 - 10 | 1406 | 80 | |
GenBank mRNA | AJ250841.1 | - | 9 - 10 | 80 | ||
AK039759.1 | - | 9 - 10 | 1376 | 80 | ||
AK148262.1 | - | 9 - 10 | 1379 | 80 | ||
BC021392.1 | - | 9 - 10 | 5069005 | 80 | ||
BC021464.1 | - | 9 - 10 | 5069004 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |