Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022007.1 | NP_071290.1 | 3 - 4 | 207 | 74 | |
GenBank mRNA | AI892161.1 | - | 3 - 4 | 207 | 74 | |
AK004113.1 | - | 3 - 4 | 216 | 74 | ||
BC048737.1 | - | 3 - 4 | 220 | 74 | ||
BC061101.1 | - | 3 - 4 | 220 | 6531288 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |