Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snap29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023348.4 | NP_075837.3 | 1 - 2 | 357 | 102 | |
GenBank mRNA | AK005042.1 | - | 1 - 2 | 307 | 102 | |
AK007065.1 | - | 1 - 2 | 350 | 102 | ||
AK039203.1 | - | 1 - 2 | 326 | 102 | ||
AK147845.1 | - | 1 - 2 | 265 | 102 | ||
AY008722.1 | - | 1 - 2 | 354 | 102 | ||
BC030066.1 | - | 1 - 2 | 306 | 5118167 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |