Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tinagl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168333.1 | NP_001161805.1 | 10 - 11 | 1300 | 60 | |
NM_023476.3 | NP_075965.2 | 10 - 11 | 1441 | 60 | ||
GenBank mRNA | AB047402.1 | - | 10 - 11 | 1442 | 60 | |
AB050626.1 | - | 10 - 11 | 1278 | 60 | ||
BC005738.1 | - | 10 - 11 | 1268 | 2650851 | 60 | |
BC018539.1 | - | 10 - 11 | 1285 | 3596560 | 60 | |
CT010275.1 | - | 9 - 10 | 1059 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |