Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023733.3 | NP_076222.1 | 10 - 11 | 1134 | 122 | |
GenBank mRNA | AK004567.2 | - | 10 - 11 | 1135 | 122 | |
BC006593.1 | - | 10 - 11 | 1122 | 3156501 | 122 | |
BC012308.1 | - | 10 - 11 | 1149 | 3500167 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |