Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023844.5 | NP_076333.3 | 5 - 6 | 961 | 93 | |
GenBank mRNA | AF255911.1 | - | 5 - 6 | 684 | 93 | |
AJ291757.1 | - | 5 - 6 | 93 | |||
AK010616.1 | - | 5 - 6 | 803 | 93 | ||
AK013914.1 | - | 5 - 6 | 802 | 93 | ||
AK028757.1 | - | 5 - 6 | 652 | 93 | ||
AK078128.1 | - | 5 - 6 | 687 | 93 | ||
AK153636.1 | - | 5 - 6 | 652 | 93 | ||
BC028778.1 | - | 5 - 6 | 961 | 1195543 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |