Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024258.2 | NP_077220.2 | 6 - 7 | 789 | 94 | |
GenBank mRNA | AK004594.2 | - | 6 - 7 | 764 | 94 | |
AK018212.1 | - | 6 - 7 | 722 | 94 | ||
AK160038.1 | - | 6 - 7 | 759 | 94 | ||
BC003278.1 | - | 6 - 7 | 718 | 3582180 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |