Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026175.5 | NP_080451.4 | 3 - 4 | 532 | 70 | |
GenBank mRNA | AI115048.1 | - | 3 - 4 | 533 | 70 | |
AK004763.1 | - | 3 - 4 | 515 | 70 | ||
AK028829.1 | - | 3 - 4 | 484 | 70 | ||
AK029095.1 | - | 3 - 4 | 502 | 70 | ||
AK030223.1 | - | 3 - 4 | 519 | 70 | ||
AK051703.1 | - | 3 - 4 | 532 | 70 | ||
AK160061.1 | - | 3 - 4 | 515 | 70 | ||
AK202411.1 | - | 1 - 2 | 116 | 70 | ||
AK210145.1 | - | 1 - 2 | 116 | 70 | ||
BC029753.1 | - | 3 - 4 | 496 | 2651773 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |