Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ero1lb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026184.2 | NP_080460.2 | 11 - 12 | 1003 | 95 | |
GenBank mRNA | AF489856.1 | - | 11 - 12 | 809 | 95 | |
AK004981.1 | - | 980 | 95 | |||
AK041951.1 | - | 11 - 12 | 979 | 95 | ||
AK160664.1 | - | 11 - 12 | 1019 | 95 | ||
BC058721.1 | - | 11 - 12 | 1003 | 5705507 | 95 | |
BC111900.1 | - | 11 - 12 | 973 | 40062137 | 95 | |
BC112426.1 | - | 11 - 12 | 973 | 40062136 | 95 | |
BC145888.1 | - | 11 - 12 | 954 | 40131191 | 95 | |
BC145890.1 | - | 11 - 12 | 954 | 40131193 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |