Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026232.3 | NP_080508.1 | 8 - 9 | 930 | 103 | |
GenBank mRNA | AK015679.1 | - | 8 - 9 | 931 | 103 | |
AK017035.1 | - | 8 - 9 | 931 | 103 | ||
AK030950.1 | - | 8 - 9 | 899 | 103 | ||
AK090086.1 | - | 8 - 9 | 867 | 103 | ||
AK151334.1 | - | 8 - 9 | 899 | 103 | ||
AK151420.1 | - | 8 - 9 | 867 | 103 | ||
AK159122.1 | - | 8 - 9 | 928 | 103 | ||
AK159732.1 | - | 8 - 9 | 928 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |