Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026395.1 | NP_080671.1 | 3 - 4 | 280 | 71 | |
GenBank mRNA | AK002550.1 | - | 3 - 4 | 280 | 71 | |
AK003682.1 | - | 3 - 4 | 302 | 71 | ||
AK013262.1 | - | 3 - 4 | 337 | 71 | ||
AK078396.1 | - | 3 - 4 | 343 | 71 | ||
AK168192.1 | - | 4 - 5 | 499 | 71 | ||
BC029189.1 | - | 2 - 3 | 186 | 4036721 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |