Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026403.3 | NP_080679.3 | 2 - 3 | 777 | 95 | |
GenBank mRNA | AK011570.1 | - | 2 - 3 | 767 | 95 | |
AK015585.1 | - | 2 - 3 | 527 | 95 | ||
AK032873.1 | - | 2 - 3 | 761 | 95 | ||
AK037831.1 | - | 2 - 3 | 765 | 95 | ||
AK050912.1 | - | 2 - 3 | 768 | 96 | ||
AK083861.1 | - | 2 - 3 | 646 | 95 | ||
AK139689.1 | - | 2 - 3 | 728 | 95 | ||
AK146020.1 | - | 2 - 3 | 732 | 95 | ||
AK146687.1 | - | 2 - 3 | 749 | 95 | ||
AK152732.1 | - | 2 - 3 | 751 | 95 | ||
AK154387.1 | - | 2 - 3 | 769 | 95 | ||
AK159798.1 | - | 2 - 3 | 751 | 95 | ||
BC018523.1 | - | 2 - 3 | 757 | 3499986 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |