Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900006621.mm.1 |
NM_026635.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900006621.mm.1 |
NM_026635.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900006621.mm.1 |
TC0900000863.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900006619.mm.1 |
NM_026635.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900006619.mm.1 |
NM_026635.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900006619.mm.1 |
TC0900000863.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900006623.mm.1 |
NM_026635.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900006623.mm.1 |
NM_026635.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900006623.mm.1 |
TC0900000863.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900006618.mm.1 |
NM_026635.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900006618.mm.1 |
NM_026635.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900006618.mm.1 |
TC0900000863.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900006620.mm.1 |
NM_026635.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900006620.mm.1 |
NM_026635.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900006620.mm.1 |
TC0900000863.mm.1 |
MTA TC |