Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027019.3 | NP_081295.2 | 4 - 5 | 437 | 67 | |
GenBank mRNA | AB067773.1 | - | 4 - 5 | 526 | 67 | |
AK005883.1 | - | 4 - 5 | 548 | 67 | ||
BC104105.1 | - | 4 - 5 | 437 | 40043157 | 67 | |
BC104106.1 | - | 4 - 5 | 437 | 40043160 | 67 | |
BC106806.1 | - | 4 - 5 | 437 | 40042632 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |