Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027790.2 | NP_082066.2 | 6 - 7 | 651 | 83 | |
GenBank mRNA | AK017274.1 | - | 6 - 7 | 653 | 83 | |
BC117731.2 | - | 5 - 6 | 544 | 40089799 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |