Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900008653.mm.1 |
NM_027877.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008653.mm.1 |
NM_027877.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008653.mm.1 |
TC1900000969.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900008656.mm.1 |
NM_027877.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008656.mm.1 |
NM_027877.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008656.mm.1 |
TC1900000969.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900008649.mm.1 |
NM_027877.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008649.mm.1 |
NM_027877.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008649.mm.1 |
TC1900000969.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900008655.mm.1 |
NM_027877.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008655.mm.1 |
NM_027877.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008655.mm.1 |
TC1900000969.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900008652.mm.1 |
NM_027877.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008652.mm.1 |
NM_027877.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008652.mm.1 |
TC1900000969.mm.1 |
MTA TC |