Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800002909.mm.1 |
NM_031374.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002909.mm.1 |
NM_031374.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002909.mm.1 |
TC0800000390.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800002912.mm.1 |
NM_031374.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002912.mm.1 |
NM_031374.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002912.mm.1 |
TC0800000390.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800002910.mm.1 |
NM_031374.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002910.mm.1 |
NM_031374.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002910.mm.1 |
TC0800000390.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800002911.mm.1 |
NM_031374.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002911.mm.1 |
NM_031374.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002911.mm.1 |
TC0800000390.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800002908.mm.1 |
NM_031374.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002908.mm.1 |
NM_031374.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002908.mm.1 |
TC0800000390.mm.1 |
MTA TC |