Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fstl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031380.2 | NP_113557.1 | 1 - 2 | 126 | 72 | |
GenBank mRNA | AB024429.1 | - | 1 - 2 | 99 | 72 | |
AF276238.1 | - | 1 - 2 | 105 | 72 | ||
AF310616.1 | - | 1 - 2 | 99 | 72 | ||
AK082898.1 | - | 1 - 2 | 128 | 72 | ||
AK087247.1 | - | 1 - 2 | 107 | 72 | ||
BC061052.1 | - | 1 - 2 | 126 | 30312598 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |