Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spock2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052994.2 | NP_443720.1 | 9 - 10 | 1200 | 66 | |
GenBank mRNA | AJ278999.1 | - | 9 - 10 | 1202 | 66 | |
AK122233.1 | - | 9 - 10 | 862 | 66 | ||
AK140483.1 | - | 9 - 10 | 1107 | 66 | ||
BC057324.1 | - | 9 - 10 | 1200 | 6821449 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |