Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053246.3 | NP_444476.1 | 5 - 6 | 733 | 81 | |
GenBank mRNA | AF418207.1 | - | 4 - 5 | 617 | 81 | |
AK032352.1 | - | 5 - 6 | 723 | 81 | ||
AK043670.1 | - | 4 - 5 | 645 | 81 | ||
AK077612.1 | - | 5 - 6 | 725 | 81 | ||
AK139363.1 | - | 4 - 5 | 621 | 81 | ||
AK142621.1 | - | 4 - 5 | 640 | 81 | ||
AK156485.1 | - | 5 - 6 | 733 | 81 | ||
BC004705.1 | - | 5 - 6 | 694 | 3499419 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |