Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adssl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007421.2 | NP_031447.1 | 10 - 11 | 1146 | 63 | |
GenBank mRNA | AK089520.1 | - | 10 - 11 | 1121 | 63 | |
AK146907.1 | - | 10 - 11 | 1149 | 63 | ||
AK149634.1 | - | 10 - 11 | 1121 | 63 | ||
AK150874.1 | - | 10 - 11 | 1117 | 63 | ||
BC039943.1 | - | 10 - 11 | 1177 | 4014672 | 63 | |
M74495.1 | - | 10 - 11 | 1104 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |