Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077264.1 | NP_001070732.1 | 3 - 4 | 419 | 76 | |
NM_007458.2 | NP_031484.1 | 3 - 4 | 419 | 76 | ||
GenBank mRNA | AK135424.1 | - | 3 - 4 | 348 | 76 | |
AK155515.1 | - | 3 - 4 | 355 | 76 | ||
AK208311.1 | - | 2 - 3 | 206 | 76 | ||
AK212972.1 | - | 3 - 4 | 324 | 76 | ||
BC031433.1 | - | 3 - 4 | 369 | 4481643 | 76 | |
CA327188.1 | - | 3 - 4 | 425 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |