Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007488.3 | NP_031514.3 | 1 - 2 | 179 | 77 | |
GenBank mRNA | BC054546.1 | - | 1 - 2 | 100 | 6409636 | 77 |
BU056629.1 | - | 1 - 2 | 317 | 77 | ||
D63644.1 | - | 1 - 2 | 55 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |