Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400022919.mm.1 |
TC0400002791.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400022919.mm.1 |
NM_001276446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400022919.mm.1 |
NM_008525.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400022918.mm.1 |
TC0400002791.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400022918.mm.1 |
NM_001276446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400022918.mm.1 |
NM_008525.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400022929.mm.1 |
TC0400002791.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400022929.mm.1 |
NM_008525.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400022916.mm.1 |
TC0400002791.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400022916.mm.1 |
NM_001276446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400022916.mm.1 |
NM_008525.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400022922.mm.1 |
TC0400002791.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400022922.mm.1 |
NM_001276446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400022922.mm.1 |
NM_008525.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400022923.mm.1 |
TC0400002791.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400022923.mm.1 |
NM_001276446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400022923.mm.1 |
NM_008525.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400022920.mm.1 |
TC0400002791.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400022920.mm.1 |
NM_001276446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400022920.mm.1 |
NM_008525.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400022921.mm.1 |
TC0400002791.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400022921.mm.1 |
NM_001276446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400022921.mm.1 |
NM_008525.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400022924.mm.1 |
TC0400002791.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400022924.mm.1 |
NM_001276446.1 |
Refseq |