Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000019098.mm.1 |
NM_008688.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019098.mm.1 |
NM_008688.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019098.mm.1 |
NM_026756.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019098.mm.1 |
NM_026756.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019098.mm.1 |
TC1000002541.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019094.mm.1 |
NM_008688.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019094.mm.1 |
NM_008688.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019094.mm.1 |
NM_026756.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019094.mm.1 |
NM_026756.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019094.mm.1 |
TC1000002541.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019090.mm.1 |
NM_008688.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019090.mm.1 |
NM_008688.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019090.mm.1 |
NM_026756.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019090.mm.1 |
NM_026756.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019090.mm.1 |
TC1000002541.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019092.mm.1 |
NM_008688.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019092.mm.1 |
NM_008688.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019092.mm.1 |
NM_026756.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019092.mm.1 |
NM_026756.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019092.mm.1 |
TC1000002541.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019091.mm.1 |
NM_008688.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019091.mm.1 |
NM_008688.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019091.mm.1 |
NM_026756.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019091.mm.1 |
NM_026756.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019091.mm.1 |
TC1000002541.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019093.mm.1 |
NM_008688.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019093.mm.1 |
NM_008688.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019093.mm.1 |
NM_026756.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019093.mm.1 |
NM_026756.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019093.mm.1 |
TC1000002541.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019100.mm.1 |
NM_008688.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019100.mm.1 |
NM_008688.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019100.mm.1 |
TC1000002541.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019096.mm.1 |
NM_008688.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019096.mm.1 |
NM_008688.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019096.mm.1 |
NM_026756.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019096.mm.1 |
NM_026756.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019096.mm.1 |
TC1000002541.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019095.mm.1 |
NM_008688.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019095.mm.1 |
NM_008688.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019095.mm.1 |
NM_026756.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019095.mm.1 |
NM_026756.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019095.mm.1 |
TC1000002541.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019097.mm.1 |
NM_008688.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019097.mm.1 |
NM_008688.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019097.mm.1 |
NM_026756.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019097.mm.1 |
NM_026756.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019097.mm.1 |
TC1000002541.mm.1 |
MTA TC |