Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcolce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008788.2 | NP_032814.2 | 3 - 4 | 534 | 83 | |
GenBank mRNA | AB008548.1 | - | 3 - 4 | 458 | 83 | |
AK142615.1 | - | 3 - 4 | 463 | 83 | ||
AK144392.1 | - | 3 - 4 | 535 | 83 | ||
AK146829.1 | - | 3 - 4 | 535 | 83 | ||
AK159430.1 | - | 3 - 4 | 522 | 83 | ||
AK159940.1 | - | 3 - 4 | 527 | 83 | ||
AK162524.1 | - | 3 - 4 | 538 | 83 | ||
CX210896.1 | - | 1 - 2 | 216 | 83 | ||
X57337.1 | - | 3 - 4 | 501 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |