Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285890.1 | NP_001272819.1 | 11 - 12 | 1594 | 68 | |
NM_008800.2 | NP_032826.1 | 12 - 13 | 1611 | 68 | ||
GenBank mRNA | AK004772.1 | - | 11 - 12 | 1201 | 68 | |
BC058531.1 | - | 12 - 13 | 1611 | 6826674 | 68 | |
L01695.1 | - | 12 - 13 | 1354 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |