Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166388.1 | NP_001159860.1 | 2 - 3 | 1537 | 67 | |
NM_001166389.1 | NP_001159861.1 | 2 - 3 | 1268 | 67 | ||
NM_008975.3 | NP_033001.2 | 2 - 3 | 1337 | 67 | ||
GenBank mRNA | AF035645.1 | - | 1 - 2 | 318 | 67 | |
AK003954.1 | - | 1 - 2 | 75 | 67 | ||
AK004562.1 | - | 2 - 3 | 947 | 67 | ||
AK014601.1 | - | 2 - 3 | 951 | 67 | ||
AK143702.1 | - | 2 - 3 | 989 | 67 | ||
AK172192.1 | - | 2 - 3 | 991 | 67 | ||
BC027445.1 | - | 1 - 2 | 148 | 4482106 | 67 | |
BC066043.1 | - | 2 - 3 | 1537 | 6415021 | 67 | |
CT010215.1 | - | 1 - 2 | 103 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |